Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Phenylalanine benzyl ester p-toluenesulfonate, 98%

Click to view available options
Quantity:
5g
25g
100g
28607-46-7
C23H25NO5S
427.515
MFCD00066129
ZLZGBBIPWXUQST-XFULWGLBSA-N
h-d-phe-obzl tos,d-phenylalanine benzyl ester p-toluenesulfonate,h-d-phe-obzl.tosoh,d-phenylalanine benzyl ester tosylate,r-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate,benzyl 2r-2-amino-3-phenylpropanoate; para-toluene sulfonate,d-phe-obzl,h-d-phe-obzl.tos,h-d-phe-obzl?os,h-d-phe-obzl. tos
44629866
benzyl (2R)-2-amino-3-phenylpropanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)CC(C(=O)OCC2=CC=CC=C2)N
28607-46-7
C23H25NO5S
427.515
MFCD00066129
ZLZGBBIPWXUQST-XFULWGLBSA-N
h-d-phe-obzl tos,d-phenylalanine benzyl ester p-toluenesulfonate,h-d-phe-obzl.tosoh,d-phenylalanine benzyl ester tosylate,r-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate,benzyl 2r-2-amino-3-phenylpropanoate; para-toluene sulfonate,d-phe-obzl,h-d-phe-obzl.tos,h-d-phe-obzl?os,h-d-phe-obzl. tos
44629866
benzyl (2R)-2-amino-3-phenylpropanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)CC(C(=O)OCC2=CC=CC=C2)N
Beilstein 5375462
Formula Weight 427.51
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material D-Phenylalanine benzyl ester p-toluenesulfonate

storageNote1 : 2° to 8°C