Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Phenylalaninol, 98%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
5267-64-1
C9H13NO
MFCD00064298
d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol
5267-64-1
C9H13NO
MFCD00064298
d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

Solubility
Soluble in Dichloromethane, Ethyl Acetate, Methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with carbondioxide, air, acids and oxidizing agents.
Melting Point 89°C to 94°C
Quantity 1g
Optical Rotation +23 ± 2° (c=5 in Ethanol)
Sensitivity Air Sensitive
UN Number UN3259
Beilstein 4665408
Solubility Information Soluble in Dichloromethane,Ethyl Acetate,Methanol.
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material D-Phenylalaninol

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

EINECSNumber : 226-086-1

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

RUO – Research Use Only