Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D(-)-Phenylglycinamide, 99%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Specifications
Color | White to Yellow |
Melting Point | 125.0°C to 129.0°C |
Quantity | 5g |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Specific Rotation | − 48.50 |
Specific Rotation Condition | −48.5° (20°C c=0.55,EtOH) |
Assay Percent Range | 98.5% min. (GC) |
Linear Formula | C6H5CH(NH2)C(O)NH2 |
Solubility Information | Solubility in water: soluble. Other solubilities: better soluble ph <6 |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning