Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Pipecolinic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
1723-00-8
C6H11NO2
129.159
MFCD00064346
HXEACLLIILLPRG-RXMQYKEDSA-N
r-piperidine-2-carboxylic acid,d-pipecolic acid,d-pipecolinic acid,d +-pipecolinic acid,d-homoproline,2r-piperidine-2-carboxylic acid,r-pipecolic acid,+-pipecolic acid,d-piperidine-2-carboxylic acid
736316
CHEBI:41582
(2R)-piperidine-2-carboxylic acid
C1CCNC(C1)C(=O)O
1723-00-8
C6H11NO2
129.159
MFCD00064346
HXEACLLIILLPRG-RXMQYKEDSA-N
r-piperidine-2-carboxylic acid,d-pipecolic acid,d-pipecolinic acid,d +-pipecolinic acid,d-homoproline,2r-piperidine-2-carboxylic acid,r-pipecolic acid,+-pipecolic acid,d-piperidine-2-carboxylic acid
736316
CHEBI:41582
(2R)-piperidine-2-carboxylic acid
C1CCNC(C1)C(=O)O
Melting Point 277°C (decomposition)
Quantity 5g
Beilstein 81094
Formula Weight 129.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material D-Pipecolinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-024-4

TSCA : No

Recommended Storage : Ambient temperatures