Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Prolinamide hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
50894-62-7
C5H11ClN2O
150.606
MFCD00237235
CSKSDAVTCKIENY-PGMHMLKASA-N
r-pyrrolidine-2-carboxamide hydrochloride,h-d-pro-nh2 hcl,d-prolinamide hydrochloride,d-proline amide hydrochloride,d-prolinamide hcl,d-prolinamide hydrochloride,r-prolinamide hydrochloride,2r-pyrrolidine-2-carboxamide hydrochloride,d-prolin amide hydrochloride,h-d-pro-nh hcl
46735192
(2R)-pyrrolidine-2-carboxamide;hydrochloride
C1CC(NC1)C(=O)N.Cl
50894-62-7
C5H11ClN2O
150.606
MFCD00237235
CSKSDAVTCKIENY-PGMHMLKASA-N
r-pyrrolidine-2-carboxamide hydrochloride,h-d-pro-nh2 hcl,d-prolinamide hydrochloride,d-proline amide hydrochloride,d-prolinamide hcl,d-prolinamide hydrochloride,r-prolinamide hydrochloride,2r-pyrrolidine-2-carboxamide hydrochloride,d-prolin amide hydrochloride,h-d-pro-nh hcl
46735192
(2R)-pyrrolidine-2-carboxamide;hydrochloride
C1CC(NC1)C(=O)N.Cl
Beilstein 6280978
Formula Weight 150.61
Physical Form Solid
Percent Purity 97%
Quantity 25g
Chemical Name or Material D-Prolinamide hydrochloride