Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Proline benzyl ester hydrochloride, 97%

Click to view available options
Quantity:
1g
5g
53843-90-6
C12H16ClNO2
241.715
MFCD00153458
NEDMOHHWRPHBAL-RFVHGSKJSA-N
h-d-pro-obzl hcl,h-d-pro-obzl.hcl,d-proline benzyl ester hydrochloride,d-proline, phenylmethyl ester, hydrochloride,r-benzyl pyrrolidine-2-carboxylate hydrochloride,benzyl 2r-pyrrolidine-2-carboxylate hydrochloride,h-d-pro-obzl-hcl,h-d-pro-obzl inverted exclamation mark currencyhcl,d-proline benzylester hydrochloride,d-proline-benzyl-ester hydrochloride
22827650
benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
53843-90-6
C12H16ClNO2
241.715
MFCD00153458
NEDMOHHWRPHBAL-RFVHGSKJSA-N
h-d-pro-obzl hcl,h-d-pro-obzl.hcl,d-proline benzyl ester hydrochloride,d-proline, phenylmethyl ester, hydrochloride,r-benzyl pyrrolidine-2-carboxylate hydrochloride,benzyl 2r-pyrrolidine-2-carboxylate hydrochloride,h-d-pro-obzl-hcl,h-d-pro-obzl inverted exclamation mark currencyhcl,d-proline benzylester hydrochloride,d-proline-benzyl-ester hydrochloride
22827650
benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
Beilstein 6852422
Formula Weight 241.71
Physical Form Solid
Percent Purity 97%
Quantity 1g
Chemical Name or Material D-Proline benzyl ester hydrochloride