Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Proline ethyl ester hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
131477-20-8
C7H14ClNO2
179.644
MFCD00191003
DUJGQVVONTYHLT-FYZOBXCZSA-N
r-ethyl pyrrolidine-2-carboxylate hydrochloride,d-proline ethyl ester hydrochloride,h-d-pro-oet hcl,h-d-pro-oet hydrochloride,d-proline, ethyl ester, hydrochloride,ethyl 2r-pyrrolidine-2-carboxylate hydrochloride,h-d-pro-oet.hcl,h-d-pro-oet . hci,h-d-pro-oet?cl,d-proline ethyl ester hcl
56777374
ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CCCN1.Cl
131477-20-8
C7H14ClNO2
179.644
MFCD00191003
DUJGQVVONTYHLT-FYZOBXCZSA-N
r-ethyl pyrrolidine-2-carboxylate hydrochloride,d-proline ethyl ester hydrochloride,h-d-pro-oet hcl,h-d-pro-oet hydrochloride,d-proline, ethyl ester, hydrochloride,ethyl 2r-pyrrolidine-2-carboxylate hydrochloride,h-d-pro-oet.hcl,h-d-pro-oet . hci,h-d-pro-oet?cl,d-proline ethyl ester hcl
56777374
ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CCCN1.Cl
Beilstein 7170607
Formula Weight 179.64
Physical Form Liquid
Percent Purity 98%
Color Yellow
Quantity 100g
Chemical Name or Material D-Proline ethyl ester hydrochloride

storageNote1 : -30° to -10°C