Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Proline Methyl Ester Hydrochloride 96.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
25g
65365-28-8
C6H12ClNO2
165.617
MFCD00083685
HQEIPVHJHZTMDP-NUBCRITNSA-N
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
44629857
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
COC(=O)C1CCCN1.Cl
65365-28-8
C6H12ClNO2
165.617
MFCD00083685
HQEIPVHJHZTMDP-NUBCRITNSA-N
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
44629857
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
COC(=O)C1CCCN1.Cl
Quantity 25g
Melting Point 73°C
Color White
Formula Weight 165.62
Physical Form Crystal-Powder at 20°C
Percent Purity ≥96.0% (N,T)
Chemical Name or Material D-Proline Methyl Ester Hydrochloride

TSCA : No

Recommended Storage : Refrigerator