Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Propargylglycine, 97%

Click to view available options
Quantity:
1g
5g
23235-03-2
C5H7NO2
113.116
MFCD00237407
DGYHPLMPMRKMPD-SCSAIBSYSA-N
d-propargylglycine,r-2-aminopent-4-ynoic acid,r-alpha-propargylglycine,unii-2252qef2pl,4-pentynoic acid, 2-amino-, 2r,2r-2-aminopent-4-ynoic acid,r-2-propargylglycine,r-2-amino-4-pentynoic acid,d-2-propynylglycine,propargylglycine, d
6992713
(2R)-2-aminopent-4-ynoic acid
C#CCC(C(=O)O)N
23235-03-2
C5H7NO2
113.116
MFCD00237407
DGYHPLMPMRKMPD-SCSAIBSYSA-N
d-propargylglycine,r-2-aminopent-4-ynoic acid,r-alpha-propargylglycine,unii-2252qef2pl,4-pentynoic acid, 2-amino-, 2r,2r-2-aminopent-4-ynoic acid,r-2-propargylglycine,r-2-amino-4-pentynoic acid,d-2-propynylglycine,propargylglycine, d
6992713
(2R)-2-aminopent-4-ynoic acid
C#CCC(C(=O)O)N
Applications
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Formula Weight 113.12
Physical Form Powder
Percent Purity 97%
Color White
Melting Point 205°C (decomposition)
Quantity 1g
Odor Odorless
Chemical Name or Material D-Propargylglycine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning