Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Salicin, 99%, Thermo Scientific Chemicals

Catalog Number aaa1652422
Click to view available options
Quantity:
5 g
25 g
100 g
138-52-3
C13H18O7
286.28
MFCD00006590
NGFMICBWJRZIBI-UJPOAAIJSA-N
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
439503
CHEBI:17814
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
138-52-3
C13H18O7
286.28
MFCD00006590
NGFMICBWJRZIBI-UJPOAAIJSA-N
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
439503
CHEBI:17814
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O

Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alkalies, pyridine, glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.

Notes
Store in cool place. Light and heat sensitive. Air senstive and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 200°C to 203°C
Assay Percent Range 99%
Quantity 100 g
Beilstein 89593
Merck Index 14,8324
Solubility Information Soluble in water,alkalies,pyridine,glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.
Formula Weight 286.28
Percent Purity 99%
Chemical Name or Material D-(-)-Salicin
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 205-331-6
RTECSNumber LZ5901700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit