Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Salicin, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
138-52-3
C13H18O7
286.27
MFCD00006590
NGFMICBWJRZIBI-UJPOAAIJSA-N
salicin,salicoside,salicine,salicyl alcohol glucoside,d---salicin,2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol,saligenin beta-d-glucopyranoside,2-hydroxymethyl phenyl beta-d-glucopyranoside,d-salicin,saligenin-beta-d-glucopyranoside
439503
CHEBI:17814
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
138-52-3
C13H18O7
286.27
MFCD00006590
NGFMICBWJRZIBI-UJPOAAIJSA-N
salicin,salicoside,salicine,salicyl alcohol glucoside,d---salicin,2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol,saligenin beta-d-glucopyranoside,2-hydroxymethyl phenyl beta-d-glucopyranoside,d-salicin,saligenin-beta-d-glucopyranoside
439503
CHEBI:17814
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
Specific Rotation -61.5
Melting Point 197°C to 200°C
Water 0.5% max.
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
Color White
Specific Rotation Condition −61.50° (20°C c=5,water)
Assay Percent Range 99+%
Beilstein 17, V,7, 113
Merck Index 15, 8460
Solubility Information Solubility in water: 36g/L (15°C),250g/L (60°C). Other solubilities: insoluble in ether
Formula Weight 286.27
Physical Form Fine Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material D(-)-Salicin, 99+%
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-331-6

RTECSNumber : LZ5901700

TSCA : TSCA