Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ D-Sorbitol, 91.0 to 100.5% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
50-70-4
C6H14O6
182.172
MFCD00004708
FBPFZTCFMRRESA-JGWLITMVSA-N
d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol
5780
CHEBI:17924
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O
50-70-4
C6H14O6
182.172
MFCD00004708
FBPFZTCFMRRESA-JGWLITMVSA-N
d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol
5780
CHEBI:17924
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O
Packaging Plastic bottle
Melting Point 98°C to 100°C
Quantity 500g
Formula Weight 182.17
Physical Form Crystalline Powder
Percent Purity 91.0 to 100.5%
Chemical Name or Material D-Sorbitol
Assay Dry basis