Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Sucrose, 99.7%, for biochemistry, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
25kg
Packaging:
Plastic bag in resealable cardbox
Plastic bottle
Plastic drum
57-50-1
C12H22O11
342.297
MFCD00006626
CZMRCDWAGMRECN-UGDNZRGBSA-N
sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
57-50-1
C12H22O11
342.297
MFCD00006626
CZMRCDWAGMRECN-UGDNZRGBSA-N
sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
Sulfated Ash 0.02 % max.
Infrared Spectrum Authentic
Loss on Drying 0.1 % max.
Color Colorless
Packaging Plastic bag in resealable cardbox
Quantity 25kg
Specific Rotation (20°C, 589nm) (c=26, H2O) > +65.9°
pH 6.5 to 7.5 (10% soln.)
Water 0.06% max.
Assay Percent Range 99.7%
Solubility Information 0.0007 % max. Insoluble in Water
Formula Weight 342.29
Physical Form Liquid
Percent Purity ≥99.7% (HPLC)
Grade Biochemical
Chemical Name or Material D(+)-Sucrose
Show More Show Less