Alfa Aesar™ D-(-)-Tartaric acid, 99%

Manufacturer: Alfa Aesar™ A1126422

Quantity 100g

 View more versions of this product

Catalog No. AAA1126422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 147-71-7
    Molecular Formula C4H6O6
    Formula Weight 150.09
    MDL Number MFCD00004238
    Beilstein 1725145
    Merck Index 14,9068
    Chemical Name or Material D- (-)-Tartaric acid

    Specifications

    Formula Weight 150.09
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Melting Point 166° to 170°C
    Quantity 100g
    Optical Rotation −13° (c=10 in water)
    Molecular Formula C4H6O6
    Merck Index 14,9068
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    pH 1.6
    Flash Point 210°C (410°F)
    MDL Number MFCD00004238
    CAS 147-71-7
    Beilstein 1725145
    Health Hazard 1 Warning
    Chemical Name or Material D- (-)-Tartaric acid
    Assay 99%

    Description

    Applications
    D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

    Solubility
    Soluble in water and alcohol.

    Notes
    Light sensitive. Incompatible with bases, oxidizing agents and reducing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit