Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
87-69-4
C4H6O6
150.086
FEWJPZIEWOKRBE-JCYAYHJZSA-N
l-tartaric acid,l-+-tartaric acid,l +-tartaric acid,2r,3r-2,3-dihydroxysuccinic acid,tartaric acid,2r,3r-2,3-dihydroxybutanedioic acid,r,r-tartaric acid,+-l-tartaric acid,dextrotartaric acid,l-threaric acid
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
C(C(C(=O)O)O)(C(=O)O)O
87-69-4
C4H6O6
150.086
FEWJPZIEWOKRBE-JCYAYHJZSA-N
l-tartaric acid,l-+-tartaric acid,l +-tartaric acid,2r,3r-2,3-dihydroxysuccinic acid,tartaric acid,2r,3r-2,3-dihydroxybutanedioic acid,r,r-tartaric acid,+-l-tartaric acid,dextrotartaric acid,l-threaric acid
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
C(C(C(=O)O)O)(C(=O)O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 150.09
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.76kg
Quantity 500g
Chemical Name or Material d-Tartaric Acid