Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

87-69-4
C4H6O6
150.086
FEWJPZIEWOKRBE-JCYAYHJZSA-N
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
C(C(C(=O)O)O)(C(=O)O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 150.09
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.76kg
Quantity 2.5kg
Chemical Name or Material d-Tartaric Acid
Grade Baker Analyzed™ ACS Reagent