Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-tert-leucine, Acros Organics

Click to view available options
Quantity:
100mg
500mg
26782-71-8
C6H13NO2
131.175
MFCD00004265
NPDBDJFLKKQMCM-BYPYZUCNSA-N
d-tert-leucine,h-d-tle-oh,d-tert-butylglycine,r-2-amino-3,3-dimethylbutanoic acid,d-t-leucine,r-tert-leucine,2r-2-amino-3,3-dimethylbutanoic acid,d-t-butylglycine,h-tbu-d-gly-oh,d-valine, 3-methyl
6950340
(2R)-2-amino-3,3-dimethylbutanoic acid
CC(C)(C)C(C(=O)O)N
26782-71-8
C6H13NO2
131.175
MFCD00004265
NPDBDJFLKKQMCM-BYPYZUCNSA-N
d-tert-leucine,h-d-tle-oh,d-tert-butylglycine,r-2-amino-3,3-dimethylbutanoic acid,d-t-leucine,r-tert-leucine,2r-2-amino-3,3-dimethylbutanoic acid,d-t-butylglycine,h-tbu-d-gly-oh,d-valine, 3-methyl
6950340
(2R)-2-amino-3,3-dimethylbutanoic acid
CC(C)(C)C(C(=O)O)N
Linear Formula (CH3)3CCH(NH2)COOH
Infrared Spectrum Authentic
Formula Weight 131.17
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Color White
Quantity 500mg
Chemical Name or Material D-tert-Leucine