Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Trehalose, Dihydrate, cGMP Manufactured, 97-102.%, Spectrum™

Click to view available options
Quantity:
1 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6138-23-4
C12H26O13
378.33
DPVHGFAJLZWDOC-PVXXTIHASA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
6138-23-4
C12H26O13
378.33
DPVHGFAJLZWDOC-PVXXTIHASA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

T1724, 6138-23-4, C12H22O11•2H2O

Spectrum™ D-(+)-Trehalose, Dihydrate, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Sulfated Ash 0.001
Quantity 25 kg
Specific Rotation +197 to +201°
CAS Max % 1
Percent Purity 97 to 102.%