Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Tryptophanol oxalate, 97%, Acros Organics

Click to view available options
Quantity:
1g
58889-66-0
C11H14N2O·C2H2O4
MFCD02094743
2r-1-hydroxy-3-1h-indol-3-yl propan-2-aminium,2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl azanium
58889-66-0
C11H14N2O·C2H2O4
MFCD02094743
2r-1-hydroxy-3-1h-indol-3-yl propan-2-aminium,2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl azanium
Specific Rotation +19° to +26° (20°C, 589nm) (c=1, H2O)
Melting Point 205°C to 210°C
Color White
Infrared Spectrum Authentic
Specific Rotation Condition +19° (20°C c=1,H2O)
Quantity 1g
Assay Percent Range 97%
Formula Weight 280.28
Physical Form Glistening Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material D-Tryptophanol oxalate
Show More Show Less