Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Tyrosinol hydrochloride, 97%, Acros Organics

Click to view available options
Quantity:
250mg
40829-04-7
C9H14ClNO2
203.666
MFCD01863503
PNGCRFWYSRUQTB-DDWIOCJRSA-N
d-tyrosinol hydrochloride,r-4-2-amino-3-hydroxypropyl phenol hydrochloride,4-2r-2-amino-3-hydroxypropyl phenol hydrochloride,2r-2-amino-3-4-hydroxyphenyl propan-1-ol, chloride
16213128
4-[(2R)-2-amino-3-hydroxypropyl]phenol;hydrochloride
C1=CC(=CC=C1CC(CO)N)O.Cl
40829-04-7
C9H14ClNO2
203.666
MFCD01863503
PNGCRFWYSRUQTB-DDWIOCJRSA-N
d-tyrosinol hydrochloride,r-4-2-amino-3-hydroxypropyl phenol hydrochloride,4-2r-2-amino-3-hydroxypropyl phenol hydrochloride,2r-2-amino-3-4-hydroxyphenyl propan-1-ol, chloride
16213128
4-[(2R)-2-amino-3-hydroxypropyl]phenol;hydrochloride
C1=CC(=CC=C1CC(CO)N)O.Cl
Specific Rotation Condition +15° (22°C C=1,H2O)
Specific Rotation +15° to +19° (20°C, 589nm) (c=1, H2O)
Color White
Infrared Spectrum Authentic
Quantity 250mg
Assay Percent Range 97%
Formula Weight 203.67
Physical Form Granular Powder
Percent Purity ≥96.0%
Chemical Name or Material D-Tyrosinol hydrochloride

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.