Alfa Aesar™ D-Valine methyl ester hydrochloride, 99%

Manufacturer: Alfa Aesar™ H2688306

Quantity 5g

 View more versions of this product

Catalog No. AAH2688306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7146-15-8
    Molecular Formula C6H13NO2·HCl
    Formula Weight 167.64
    MDL Number MFCD00237309
    Beilstein 3912091
    Synonym H-D-Val-OMe·HCl
    Chemical Name or Material D-Valine methyl ester hydrochloride

    Specifications

    Formula Weight 167.64
    Molecular Formula C6H13NO2·HCl
    MDL Number MFCD00237309
    CAS 7146-15-8
    Beilstein 3912091
    Synonym H-D-Val-OMe·HCl
    Melting Point ≈170°C
    Quantity 5g
    Recommended Storage 2° to 8°C
    Chemical Name or Material D-Valine methyl ester hydrochloride
    Assay 99%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit