Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Valine methyl ester hydrochloride, 99%

Manufacturer:  Alfa Aesar™ H2688303

Quantity 1g

 View more versions of this product

Catalog No. AAH2688303


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7146-15-8
167.64
3912091
D-Valine methyl ester hydrochloride
C6H13NO2·HCl
MFCD00237309
H-D-Val-Ome·HCl

Specifications

7146-15-8
167.64
3912091
≈170°C
D-Valine methyl ester hydrochloride
C6H13NO2·HCl
MFCD00237309
H-D-Val-Ome·HCl
1g
99%

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit