Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

d-Vitamin E Succinate, USP, 96-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
4345-03-3
C33H54O5
530.79
IELOKBJPULMYRW-NJQVLOCASA-N
4-oxo-4-{[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl]oxy}butanoic acid
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(=O)CCC(O)=O)=C(C)C(C)=C2O1
4345-03-3
C33H54O5
530.79
IELOKBJPULMYRW-NJQVLOCASA-N
4-oxo-4-{[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl]oxy}butanoic acid
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(=O)CCC(O)=O)=C(C)C(C)=C2O1

V1035, 4345-03-3, C33H54O5

Spectrum™ d-Vitamin E Succinate, USP is a fat-soluble antioxidant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 1
Percent Purity 96 to 102%