Learn More
Penta Manufacturing CoSupplier Diversity Partner D-Xylose USP, 58-86-6, MFCD00151475, 25 G DFS Item
Manufacturer:  Penta Manufacturing CoSupplier Diversity Partner 2448005
Molecular Formula: C5 H10 O5, Molecular Weight: 150.129. Xylose is a five-carbon sugar that contributes to lignocellulose in plants. Xylose is predominantly found in hardwoods and agricultural residues. D-(+)-Xylose has been used: as one of the component to stimulate microbial activity, for the enhancement of biofilm formation, as a part of biochemical tests for the identification of B. cereus group, and for substrate screening characterization.
Catalog No. NC0183712