Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ N-Desmethyldoxepin-D3 Hydrochloride Solution, cis/trans, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco D0751ML
Description
N-Desmethyldoxepin or nordoxepin is the primary metabolite in urine and plasma of doxepin, a tricyclic antidepressant marketed as Apona, Adapine, or Doxal. This internal standard is suitable for numerous GC/MS and LC/MS applications from forensic analysis and clinical toxicology testing to urine drug testing, pain prescription monitoring and isotope dilution methods.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
304.83 | |
1 mL |
C18D3H16NO · HCl | |
N-Desmethyldoxepin-D3 hydrochloride solution | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370-H412
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction