Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Docosapentaenoic acid (cis-7,10,13,16,19), 500 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco D1201ML
Description
A Certified Spiking Solution™ suitable for use in mass spectrometry-based fatty acid testing applications such as assessment of cardiovascular disease risk and fatty acid defficiency, and detection and quantification of docosapentaenoic acid in nutraceuticals and dietary supplements. Docosapentaenoic acid (cis-7,10,13,16,19), also known as DPA, is an omega-3 fatty acid associated with neurological and ocular health benefits. Studies suggests that DPA as well as other fatty acids can serve as biomarkers for cardiovascular disease, and nutritional and metabolic disorders.Specifications
64-17-5, 24880-45-3 | |
MFCD00156103 | |
500 μg/mL in ethanol | |
Ampule | |
14°C (57.2°F) - closed cup |
C22H34O2 | |
UN 1170 - class 3 - PG 2 - Ethanol | |
330.50 | |
1 mL |
Safety and Handling
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction