Learn More
Epredia™ Lab Vision™ D4-GDI Rabbit Polyclonal Antibody with BSA and azide
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ D4-GDI, Rabbit Polyclonal Antibody.
Supplier: Epredia™ RB9042R1
Description
D4-GDI belongs to the family of Rho-GDIs or GDP-dissociation inhibitors for the Rho/Rac family of small G proteins; consisting of RhoGDI-1, D4-GDI/RhoGDI-2 and RhoGDI-3. It inhibits the release of GDP from the Rho proteins, essential for GTP-loading and activation of GTPase activity. D4-GDI is located in the cytoplasm and in involved in the regulation of the membrane association and dissociation cycle of Rho/Rac proteins. It is known to be a substrate for caspase 3 during Fas-mediated apoptosis.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant full-length human D4-GDI protein
Molecular Weight: 28 kDa
Positive Control: Placenta and tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
D4-GDI | |
Polyclonal | |
Rabbit | |
1 mL | |
Apoptosis | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
Recombinant full-length human D4-GDI protein | |
RUO | |
Primary | |
Prediluted |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.