Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D4-GDI/RhoGDI2, Mouse anti-Human, Clone: 10D774, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280690
Description
D4-GDI/RhoGDI2 Monoclonal antibody specifically detects D4-GDI/RhoGDI2 in Human Samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence.Specifications
D4-GDI/RhoGDI2 | |
10D774 | |
Western Blot 1-2 ug/ml, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Purified | |
ARHGDIB | |
A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen. | |
Protein G purified | |
RUO | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Cross reacts with Human. |
Flow Cytometry, Immunofluorescence, Western Blot | |
Unconjugated | |
PBS and 0.02% gelatin. | |
ARHGDIB Rho GDP dissociation inhibitor (GDI) beta, D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
Mouse | |
IgG | |
0.1 mg | |
Apoptosis, Signal Transduction | |
Primary | |
397 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title