Learn More
Aviva Systems Biology Corp DAAM1 Antibody - middle region (ARP55131_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55131_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAAM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DAAM1. Alternate names: FLJ41657, KIAA0666ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61426