Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAAM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582927
Description
Immunogen sequence: QNDKGQDPDS TPLENFNIKN VVRMLVNENE VKQWKEQAEK MRKEHNELQQ KLEKKERECD AKTQEKEEMM QTLNKMKEKL EKETTEHKQV KQQVADLTAQ LHELSRRAVC ASIPG.
DAAM1 is a diaphanous-related formin first studied as a novel Disheveled binding protein and shown to be crucial for the planar cell polarity pathway in Xenopus. DAAM1, like other formins, directs nucleation and elongation of new actin filaments using its conserved formin-homology-2 (FH2) domain. It is a Rho-regulated formin implicated in neuronal development and regulation of gastrulation. It was identified as a binding protein of the Wnt receptor-associated protein Disheveled. It also binds the GTP-bound RhoA and increases its GTP-bound form population in cells. It exists in an autoinhibited state via intramolecular interactions between its amino-terminal GBD and carboxyl-terminal DAD domains. Structurally the protein includes a GTPase-binding domain (GBD), a diaphanous inhibitory domain (DID), and a coiled-coil region followed by the FH1, FH2 and C-terminal diaphanous auto-regulatory domain (DAD). In humans, the DAAM1 gene is mapped to chromosome 14q23.1.
Specifications
DAAM1 | |
Polyclonal | |
Unconjugated | |
DAAM1 | |
Daam1; Disheveled-associated activator of morphogenesis 1; dishevelled associated activator of morphogenesis 1; I79_021650; KIAA0666 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4D1 | |
DAAM1 | |
Recombinant protein corresponding to Human DAAM1. Recombinant protein control fragment (Product #RP-89125). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction