Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DALRD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | DALRD3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DALRD3 Polyclonal specifically detects DALRD3 in Human samples. It is validated for Western Blot.Specifications
DALRD3 | |
Polyclonal | |
Rabbit | |
Q5D0E6 | |
55152 | |
Synthetic peptides corresponding to DALRD3(DALR anticodon binding domain containing 3) The peptide sequence was selected from the middle region of DALRD3. Peptide sequence TEIFGVLSVATIKFEMLSTAPQSQLFLALADSSISTKGTKSGTFVMYNCA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DALR anticodon binding domain containing 3, DALR anticodon-binding domain-containing protein 3, FLJ10496 | |
DALRD3 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title