Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daminozide, Tocris Bioscience™

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
1596-84-5
C6H12N2O3
160.173
NOQGZXFMHARMLW-UHFFFAOYSA-N
daminozide,kylar,alar,dmasa,aminozide,n-dimethylamino succinamic acid,sadh,b-nine,dimas,succinic acid 2,2-dimethylhydrazide
15331
CHEBI:4312
4-(2,2-dimethylhydrazinyl)-4-oxobutanoic acid
CN(C)NC(=O)CCC(=O)O
This item is not returnable. View return policy
1596-84-5
C6H12N2O3
160.173
NOQGZXFMHARMLW-UHFFFAOYSA-N
daminozide,kylar,alar,dmasa,aminozide,n-dimethylamino succinamic acid,sadh,b-nine,dimas,succinic acid 2,2-dimethylhydrazide
15331
CHEBI:4312
4-(2,2-dimethylhydrazinyl)-4-oxobutanoic acid
CN(C)NC(=O)CCC(=O)O

Selective KDM2/7 inhibitor

Histone demethylase KDM2/7 subfamily inhibitor (IC50 values are 0.55, 1.5 and 2.1 μM for PHF8, KDM2A and KDM7A respectively). Exhibits ≥ 60-fold selectivity for KDM2/7 demethylases over other histone demethylases and 2-oxoglurate oxygenases (IC50 > 100 μM).
Quantity 50mg
Formula Weight 160.17
Percent Purity >98%
Chemical Name or Material Daminozide

Recommended Storage : Store at Room Temperature