Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAND5 Mouse anti-Human, Clone: OTI3A3, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503713
Description
This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to bind Nodal and to inhibit the Nodal signaling pathway which patterns left/right body asymmetry.Specifications
DAND5 | |
Monoclonal | |
Unconjugated | |
DAND5 | |
Liquid | |
DAND5 | |
Human recombit protein fragment corresponding to amino acids 23-189 of human DAND5 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI3A3 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N907 | |
CER2, CERL2, CKTSF1B3, COCO, CRL2, DANTE, GREM3, SP1 | |
Mouse | |
IgG2a | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
199699 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title