Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansyl Chloride 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
605-65-2
C12H12ClNO2S
269.743
MFCD00003985
XPDXVDYUQZHFPV-UHFFFAOYSA-N
5-Dimethylamino-1-naphthalenesulfonyl chloride
11801
CHEBI:51907
5-(dimethylamino)naphthalene-1-sulfonyl chloride
CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)Cl
605-65-2
C12H12ClNO2S
269.743
MFCD00003985
XPDXVDYUQZHFPV-UHFFFAOYSA-N
5-Dimethylamino-1-naphthalenesulfonyl chloride
11801
CHEBI:51907
5-(dimethylamino)naphthalene-1-sulfonyl chloride
CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)Cl
Assay Percent Range 98%
Infrared Spectrum Authentic
Formula Weight 269.75
Physical Form Powder, Needles or Crystals
Percent Purity 98%
Packaging Glass bottle
Solubility Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone, pyridine, benzene and dioxane
Melting Point 71.0°C to 74.0°C
Color Beige-Brown or Gray to White
Quantity 1g
Chemical Name or Material Dansyl chloride, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-092-6

RTECSNumber : QK3688000

TSCA : TSCA