Learn More
Tocris Bioscience™ Dansyl-NECA
Potent and selective fluorescent adenosine A1 agonist
Manufacturer:  Tocris Bioscience™ 5122/1
Catalog No. 51-221
Add to cart
This item is not returnable. View return policy
Description
Description
High affinity and selective fluorescent adenosine A1 receptor agonist (Ki values are 27, 3600 and 4300 nM for A1, A3 and A2 receptors, respectively). Permits visualization of A1 receptors in the rat cerebellar cortex.
Specifications
Specifications
Dansyl-NECA
>99
1mg
Dansyl-NECA
640.75