Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansylamide, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1431-39-6
C12H14N2O2S
250.316
MFCD00004000
TYNBFJJKZPTRKS-UHFFFAOYSA-N
dansylamide,dansyl amide,5-dimethylamino naphthalene-1-sulfonamide,5-dimethylamino-1-naphthalenesulfonamide,1-naphthalenesulfonamide, 5-dimethylamino,1okl,5-dimethylamino naphthalene-1-sulphonamide,d0s7nh
65077
5-(dimethylamino)naphthalene-1-sulfonamide
CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)N
1431-39-6
C12H14N2O2S
250.316
MFCD00004000
TYNBFJJKZPTRKS-UHFFFAOYSA-N
dansylamide,dansyl amide,5-dimethylamino naphthalene-1-sulfonamide,5-dimethylamino-1-naphthalenesulfonamide,1-naphthalenesulfonamide, 5-dimethylamino,1okl,5-dimethylamino naphthalene-1-sulphonamide,d0s7nh
65077
5-(dimethylamino)naphthalene-1-sulfonamide
CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)N
Infrared Spectrum Authentic
Melting Point 217°C to 221°C
Packaging Glass bottle
Color White to Yellow
Quantity 1g
Assay Percent Range 99%
Linear Formula (CH3)2NC10H6SO2NH2
Beilstein 14, IV, 2801
Formula Weight 250.31
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Dansylamide
Show More Show Less