Learn More
Selleck Chemical LLCSupplier Diversity Partner Danusertib (PHA-739358) 10mg 827318-97-8  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S110710MG
Danusertib (PHA-739358) is an Aurora kinase inhibitor for Aurora A/B/C with IC50 of 13 nM/79 nM/61 nM in cell-free assays, modestly potent to Abl, TrkA, c-RET and FGFR1, and less potent to Lck, VEGFR2/3, c-Kit, CDK2, etc. Danusertib induces apoptosis, cell cycle arrest, and autophagy. Phase 2.
Catalog No. 50-709-9