Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP Kinase 1, p Ser308 Antibody (DKPS308) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010524
Description
DAP Kinase 1 Monoclonal specifically detects DAP Kinase 1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Microarray.Specifications
DAP Kinase 1 (p Ser308) | |
Monoclonal | |
2.0 mg/mL | |
Western Blot 1 - 2 ug/mL, ELISA, Immunoprecipitation, Microarray | |
DAPKDAP kinase 1, death-associated protein kinase 1, DKFZp781I035, EC 2.7.11, EC 2.7.11.1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
DAP Kinase 1 Phosphospecific (DKPS308) [Ser308] | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunoprecipitation | |
DKPS308 | |
Unconjugated | |
P53355 | |
DAPK1 | |
Synthetic peptide, corresponding to amino acids 303-312 (pSER308) of Human DAP Kinase 1. | |
0.05 mL | |
Apoptosis, Cancer, Phospho Specific | |
1612 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction