Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP Kinase 2 Mouse anti-Human, Rat, Canine, Monkey, PE/Atto594, Clone: OTI1C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271753PEATT594

Catalog No. N271753P594


Add to cart

Description

Description

DAP Kinase 2 Monoclonal antibody specifically detects DAP Kinase 2 in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

DAP Kinase 2
OTI1C5
Flow Cytometry
Affinity Purified, Conjugated
DAPK2
Full length recombinant protein of human DAP Kinase 2 (NP_055141) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Autophagy, Cellular Markers
Primary
23604.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1
Mouse
IgG3
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit