Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP10/HCST Mouse anti-Human, Alexa Fluor 700, Clone: 982129, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9786N100UG
This item is not returnable.
View return policy
Description
DAP10/HCST Monoclonal specifically detects DAP10/HCST in Human samples. It is validated for Flow Cytometry.Specifications
DAP10/HCST | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
DAP10PIK3AP, DKFZP586C1522, HCST, hematopoietic cell signal transducer, KAP10, KAP10DNAX-activation protein 10, kinase assoc pro of ∼10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, PIK3AP, Transmembrane adapter protein KAP10 | |
Synthetic peptide containing human DAP10/HCST Accession # Q9UBK5 | |
100 μg | |
10870 | |
Human | |
Supernatant |
Flow Cytometry | |
982129 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UBK5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human DAP10/HCST in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title