Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP12 Mouse anti-Human, Alexa Fluor™ 647, Clone: 406288, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566603
Description
DAP12 Monoclonal antibody specifically detects DAP12 in Human samples. It is validated for Flow Cytometry.Specifications
DAP12 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
DAP12 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
406288 | |
Alexa Fluor 647 | |
25 μg | |
Primary | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction