Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Tocris Bioscience™ DAPI Available on GSA/VA Contract for Federal Government customers only.
Fluorescent DNA stain
$84.70 - $351.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-481-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5748/10
10mg Each for $84.70
N/A
57-485-0 View Documents Tocris Bioscience™
5748/50
Each for $351.00
N/A
Description
Description
Fluorescent DNA stain. Binds to AT-rich regions of DNA. Cell permeable at high concentrations.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
Recommended Storage : Store at −20°C