Learn More
DAPK2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 200481AP
Catalog No. 50-173-2673
Add to cart
Description
Description
The antibody is specific to DAPK2.
This gene encodes a protein that belongs to the serine/threonine protein kinase family. This protein contains a N-terminal protein kinase domain followed by a conserved calmodulin-binding domain with significant similarity to that of death-associated protein kinase 1 (DAPK1), a positive regulator of programmed cell death. Overexpression of this gene was shown to induce cell apoptosis. It uses multiple polyadenylation sites.
Specifications
Specifications
DAPK2
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8VDF3, Q9UIK4
DAP kinase 2, DAP kinase related protein 1, DAPK2, DRP 1
Rabbit
IgG
150 μL
-20°C
Primary
13143, 23604
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
DAPK2
Liquid
DAPK2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse