Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAPP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116566
Description
Positive Samples: Rat spleen, Rat lung Immunogen sequence: SSLKDFVKHF ANQPLIGSET GTLMVLKHPY PRKVEEPSIY ESVRVHTAMQ TGRTEDDLVP TAPSLGTKEG YLTKQGGLVK TWKTRWFTLH RNELKYFKDQ MSPEPIRILD LTECSAVQFD YSQERVNCFC LVFPFRTFYL CAKTGVEADE WIKILRWKLS QIRKQLNQGE GTIR.
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
Specifications
DAPP1 | |
Polyclonal | |
Unconjugated | |
DAPP1 | |
b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26377, 27071, 362046 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9QXT1, Q9UN19 | |
DAPP1 | |
A synthetic peptide corresponding to a sequence within amino acids 100-273 of human DAPP1 (NP_0552102). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction