Learn More
Selleck Chemical LLCSupplier Diversity Partner Darapladib (SB-480848) DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S75205MG
Darapladib (SB-480848) is a reversible lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor with IC50 of 0.25 nM. Phase 3.
Catalog No. 50-194-8088