Learn More
DARC Mouse anti-Human, Alexa Fluor 594, Clone: 358307, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB4139T100UG
Catalog No. FAB4139T100
Add to cart
This item is not returnable. View return policy
Description
Description
DARC Monoclonal specifically detects DARC in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
DARC
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q16570
DARC
HEK293 human embryonic kidney cell line transfected with human DARC Met1-Ser336 Accession # Q16570
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human DARC. Stains human DARC transfected cells but not irrelevant transfectants.
Flow Cytometry
358307
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
ACKR1, CCBP1, CD234, CD234 antigen, Dfy, Duffy Antigen, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy Antigen, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPD, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Mouse
IgG2a
RUO
Primary
2532
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)