Learn More
CPC Scientific H-DArg-Pro-Lys-Pro-DPhe-Gln-DTrp-Phe-DTrp-Leu-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SUBP001A
Encompass
ONE-LETTER SEQUENCE: rPKPfQwFwLL-NH2MOLECULAR FORMULA: C79H109N19O12MOLECULAR WEIGHT:1516.86STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [96736-12-8], RESEARCH AREA: NeuropeptidesREFERENCES: B.Holst et al., Mol. Endocrinol., 17, 2201 (2003); D.A.Jones et al., Peptides, 16, 777 (1995)
J.G.Reeve and N.M.Bleehen, Biochem. Biophys. Res. Commun., 199, 1313 (1994)
H.Houben and C.Denef, Peptides, 14, 109 (1993); P.J.Woll and E.Rozengurt, Proc. Natl. Acad. Sci. USA, 85, 1859 (1988); K.Tsou et al., Eur. J. Pharmacol., 110, 155 (1985)
Catalog No. 50-211-0447
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.