Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat DARPP-32 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: R&D Systems AF6259SP
This item is not returnable.
View return policy
Description
DARPP-32 Polyclonal specifically detects DARPP-32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DARPP-32 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DARPP32, PPP1R1B, protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32) | |
Goat | |
Affinity Purified | |
RUO | |
84152 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9UD71 | |
PPP1R1B | |
E. coli-derived recombinant human DARPP-32, Arg51-Ala204, Accession # Q9UD71 | |
25 μg | |
Primary | |
Detects human, mouse, and rat DARPP-32 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction