Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARPP-32 Mouse anti-Human, Biotin, Clone: 2G6D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237288B

Catalog No. NBP237288B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DARPP-32
2G6D9
Western Blot, ELISA
Purified
PPP1R1B
Purified recombinant fragment of human PPP1R1B (AA: 95-204) expressed in E. Coli.
Protein A or G purified
Breast Cancer, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Signal Transduction
Primary
84152
ELISA, Western Blot
Biotin
PBS with 0.05% Sodium Azide
protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32)
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit