Learn More
DAT1 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS5938R
Catalog No. 50-198-6921
Add to cart
Description
Description
The protein encoded by this gene belongs to the rhombotin family of cysteine-rich LIM domain oncogenes. This gene is predominantly expressed in the brain. Related family members, LMO1 and LMO2 on chromosome 11, have been reported to be involved in chromosomal translocations in T-cell leukemia. Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Aug 2011].
Specifications
Specifications
DAT1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LMO3
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
55885
LMO3
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q8TAP4
AI854781; BB106490; LIM domain only 3; LIM domain only 3 (rhombotin-like 2); LIM domain only protein 3; LIM only 3; Lmo3; LMO-3; Neuronal-specific transcription factor DAT1; RBTN3; Rbtn-3; RBTNL2; RGD1561357; RHOM3; Rhom-3; Rhombotin-3
KLH conjugated synthetic peptide derived from human DAT1/LIM domain only 3.
Protein A
RUO
Antibody
Polyclonal
Human